(4-Methylphenylthio)acetone ≥95%

Supplier: Matrix Scientific

005732-1G 005732-5G
101789-458EA 24.62 USD
101789-458 101789-460
(4-Methylphenylthio)acetone ≥95%
(4-Methylphenylthio)acetone

Hazard: Stench

Formula: C₁₀H₁₂OS
Boiling Pt: 96…98 °C (0.1 mmHg)
MDL Number: MFCD00052713
CAS Number: 1200-13-1

Order Now

Specification Test Results

Boiling point 96-98°C/0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR