Perfluoro(4-methyl-2-pentenoic acid) ≥97%

Supplier: Matrix Scientific

Synonyms: (E)-Perfluoro(4-methylpent-2-enoic acid), Hexafluoro-4-(trifluoromethyl)pent-2-enoic acid, Nonafluoro(4-methylpent-2-enoic acid), 4-(Trifluoromethyl)hexafluoropent-2-enoic acid

005468-5G 005468-25G
101789-004EA 103.08 USD
101789-004 101789-006
Perfluoro(4-methyl-2-pentenoic acid) ≥97%
Perfluoro(4-methyl-2-pentenoic acid)

Hazard: Corrosive

Formula: C₆HF₉O₂
MW: 276.06 g/mol
Boiling Pt: 173-174°C
MDL Number: MFCD00153286
CAS Number: 103229-89-6

Order Now

Specification Test Results

Boiling point 173-174°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR