Ethyl-3-cyano-4-(dimethylamino)-2-pyridinecarboxylate ≥90%

Supplier: Matrix Scientific

054517-500MG 054517-1G
101907-596EA 1109.23 USD
101907-596 101907-598
Ethyl-3-cyano-4-(dimethylamino)-2-pyridinecarboxylate ≥90%
Ethyl-3-cyano-4-(dimethylamino)-2-pyridinecarboxylate

Hazard: Irritant

Formula: C₁₁H₁₃N₃O₂
MW: 219.25 g/mol
Boiling Pt: 220°C@2mBar
MDL Number: MFCD14584812
CAS Number: 1221791-89-4

Order Now

Specification Test Results

Boiling point 220°C@2mBar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR