4-(Trifluoromethyl)-2-pyridinecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Trifluoromethyl)picolinonitrile

054477-500MG 054477-1G
101907-424EA 246.15 USD
101907-424 101907-426
4-(Trifluoromethyl)-2-pyridinecarbonitrile ≥95%
4-(Trifluoromethyl)-2-pyridinecarbonitrile

Hazard: Irritant

Formula: C₇H₃F₃N₂
MW: 172.11 g/mol
Boiling Pt: 58…60 °C (1.5 mmHg)
MDL Number: MFCD09038236
CAS Number: 936841-69-9

Order Now

Specification Test Results

Boiling point 58-60°C@1.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR