1-Bromo-3-(2-bromoethoxy)benzene ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(2-Bromoethoxy)bromobenzene

047272-500MG 047272-5G
101879-228EA 278.46 USD
101879-228 101879-232
1-Bromo-3-(2-bromoethoxy)benzene ≥95%
1-Bromo-3-(2-bromoethoxy)benzene

Hazard: Irritant

Formula: C₈H₈Br₂O
MW: 279.97 g/mol
Boiling Pt: 125 °C (1.7 mmHg)
MDL Number: MFCD03964543
CAS Number: 18800-29-8

Order Now

Specification Test Results

Boiling point 125°C/1.7mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR