Ethyl-2-(1-benzyl-4-piperidinylidene)acetate ≥95%

Supplier: Matrix Scientific

047072-500MG 047072-1G 047072-5G
101878-326EA 150.77 USD
101878-326 101878-328 101878-330
Ethyl-2-(1-benzyl-4-piperidinylidene)acetate ≥95%
Ethyl-2-(1-benzyl-4-piperidinylidene)acetate

Hazard: Irritant

Formula: C₁₆H₂₁NO₂
Boiling Pt: 130 °C (1 mmHg)
MDL Number: MFCD08437068
CAS Number: 40110-55-2

Order Now

Specification Test Results

Boiling point 130°C/1mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR