1-Benzyl-N-methyl-4-piperidinamine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Methylamino)-1-benzylpiperidine

046750-500MG 046750-1G 046750-5G
101876-856EA 278.46 USD
101876-856 101876-858 101876-860
1-Benzyl-N-methyl-4-piperidinamine ≥95%
1-Benzyl-N-methyl-4-piperidinamine

Hazard: Irritant

Formula: C₁₃H₂₀N₂
MW: 204.32 g/mol
Boiling Pt: 110°C/0.7mm
MDL Number: MFCD03931054
CAS Number: 7006-50-0

Order Now

Specification Test Results

Boiling point 110°C/0.7mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR