7-(Trifluoromethyl)-1,2,3,4-tetrahydroquinoline ≥97%
Supplier: Matrix Scientific
Synonyms:
1,2,3,4-Tetrahydro-7-(trifluoromethyl)quinoline
023799-250MG
023799-1G
023799-5G
101816-776EA
24.62
USD
101816-776
101816-778
101816-780
7-(Trifluoromethyl)-1,2,3,4-tetrahydroquinoline ≥97%
7-(Trifluoromethyl)-1,2,3,4-tetrahydroquinoline
Hazard: Irritant
Formula:
C₁₀H₁₀F₃N MW: 201.19 Boiling Pt: 135…140 °C (22 mmHg) |
MDL Number:
MFCD00079784 CAS Number: 450-62-4 |
Specification Test Results
Boiling point | 135-140°C/2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...