2,2-Dimethyldihydro-2H-pyran-4(3H)-one ≥95%

Supplier: Matrix Scientific

Synonyms: Tetrahydro-2,2-dimethyl-4H-pyran-4-one, 2,2-Dimethyltetrahydropyran-4-one

021441-250MG 021441-1G
101812-118EA 32.31 USD
101812-118 101812-120
2,2-Dimethyldihydro-2H-pyran-4(3H)-one ≥95%
2,2-Dimethyldihydro-2H-pyran-4(3H)-one

Hazard: Irritant/Refrigerate

Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 60…64 °C (8 mmHg)
MDL Number: MFCD01549337
CAS Number: 1194-16-7

Order Now

Specification Test Results

Boiling point 60-64°C/8mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR