2,2-Difluoropropionic acid ethyl ester ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2,2-Difluoropropionate, 2,2-Difluoropropionic acid ethyl ester

021388-5G 021388-25G
101811-936EA 110.77 USD
101811-936 101811-938
2,2-Difluoropropionic acid ethyl ester ≥97%
2,2-Difluoropropionic acid ethyl ester

Hazard: Flammable

Formula: C₅H₈F₂O₂
Boiling Pt: 24 °C (20 mmHg)
Density: 1.102
MDL Number: MFCD06657970
CAS Number: 28781-85-3

Order Now

Specification Test Results

Boiling point 24°C/20mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR