3,3,3-Trifluoro-2,2-dimethylpropionic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 3,3,3-Trifluoro-2,2-dimethylpropanoic acid

020958-1G 020958-5G
101811-094EA 66.15 USD
101811-094 101811-096
3,3,3-Trifluoro-2,2-dimethylpropionic acid ≥97%
3,3,3-Trifluoro-2,2-dimethylpropionic acid

Hazard: Corrosive/Irritant

Formula: C₅H₇F₃O₂
Boiling Pt: 76…77 °C (14 mmHg)
Melting Pt: 66…73 °C
MDL Number: MFCD08445819
CAS Number: 889940-13-0

Order Now

Specification Test Results

Boiling point 76-77°C/14m


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR