Tris[(trifluoroethoxy)methane] ≥99%

Supplier: Matrix Scientific

Synonyms: Tris(2,2,2-trifluoroethyl)orthoformate

011113-5G 011113-25G
101799-892EA 158.46 USD
101799-892 101799-894
Tris[(trifluoroethoxy)methane] ≥99%
Tris[(trifluoroethoxy)methane]

Hazard: Irritant

Formula: C₇H₇F₉O₃
MW: 310.12 g/mol
Boiling Pt: 144…146 °C
MDL Number: MFCD04973415
CAS Number: 58244-27-2

Order Now

Specification Test Results

Boiling point 144-146°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR