1,2-Dichloro-3-fluorobenzene ≥99%
Supplier: Matrix Scientific
005281-5G
005281-25G
101788-584EA
47.69
USD
101788-584
101788-586
1,2-Dichloro-3-fluorobenzene ≥99%
1,2-Dichloro-3-fluorobenzene
TCA listed: T
Hazard: Irritant
Formula:
C₆H₃Cl₂F Boiling Pt: 80 °C (22 mmHg) |
MDL Number:
MFCD00060647 CAS Number: 36556-50-0 |
Specification Test Results
Boiling point | 80°C/22mm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...