1,3-Dichloro-5-fluorobenzene ≥98%

Supplier: Matrix Scientific

Synonyms: 3,5-Dichlorofluorobenzene

005280-5G 005280-25G 005280-100G
101788-578EA 26.15 USD
101788-578 101788-580 101788-582
1,3-Dichloro-5-fluorobenzene ≥98%
1,3-Dichloro-5-fluorobenzene

Hazard: Irritant

Formula: C₆H₃Cl₂F
MW: 164.9900 g/mol
Boiling Pt: 68 °C (22 mmHg)
MDL Number: MFCD00060652
CAS Number: 1435-46-7

Order Now

Specification Test Results

Boiling point 68°C/22mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR