2,2-Difluoroethylamine ≥97%

Supplier: Matrix Scientific

005275-5G 005275-25G
101788-566EA 60 USD
101788-566 101788-568
2,2-Difluoroethylamine ≥97%
2,2-Difluoroethylamine

Hazard: Corrosive

Formula: C₂H₅F₂N
Boiling Pt: 66-67°C
MDL Number: MFCD00798137
CAS Number: 430-67-1

Order Now

Specification Test Results

Boiling point 66-67°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR