Ethyl thiophene-2-glyoxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-oxo-2-(thiophen-2-yl)acetate

004924-25G 004924-100G
101788-006EA 207.69 USD
101788-006 101788-008
Ethyl thiophene-2-glyoxylate ≥97%
Ethyl thiophene-2-glyoxylate

Hazard: Irritant

Formula: C₈H₈O₃S
MW: 184.21
Boiling Pt: 102…110 °C (0.8 mmHg)
MDL Number: MFCD00015538
CAS Number: 4075-58-5

Order Now

Specification Test Results

Boiling point 102-110°C/0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR