1-(2-Bromo-4,6-difluorophenoxy)-2-chloroethane ≥97%

Supplier: Matrix Scientific

004791-1G 004791-5G
101787-814EA 106.15 USD
101787-814 101787-816
1-(2-Bromo-4,6-difluorophenoxy)-2-chloroethane ≥97%
1-(2-Bromo-4,6-difluorophenoxy)-2-chloroethane

Hazard: Irritant

Formula: C₈H₆BrClF₂O
Boiling Pt: 119…122 °C (15 mmHg)
MDL Number: MFCD00068037
CAS Number: 175203-19-7

Order Now

Specification Test Results

Boiling point 119-122°C/1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR