1,2-Dibromo-3,3,3-trifluoropropene ≥98%

Supplier: Matrix Scientific

Synonyms: 1,2-Dibromo-3,3,3-trifluoroprop-1-ene

004543-1G 004543-5G
101787-440EA 44.62 USD
101787-440 101787-442
1,2-Dibromo-3,3,3-trifluoropropene ≥98%
1,2-Dibromo-3,3,3-trifluoropropene

Hazard: Irritant

Formula: C₃HBr₂F₃
MW: 253.85 g/mol
Boiling Pt: 98…100 °C
MDL Number: MFCD00042236
CAS Number: 431-22-1

Order Now

Specification Test Results

Boiling point 98-100°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR