1,2-Dibromo-1-chloro-2,2-difluoroethane ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Chloro-1,2-dibromo-2,2-difluoroethane

004533-1G 004533-5G
101787-410EA 83.08 USD
101787-410 101787-412
1,2-Dibromo-1-chloro-2,2-difluoroethane ≥97%
1,2-Dibromo-1-chloro-2,2-difluoroethane

Hazard: Irritant

Formula: C₂HBr₂ClF₂
MW: 258.3 g/mol
Boiling Pt: 117…119 °C
MDL Number: MFCD01075262
CAS Number: 421-36-3

Order Now

Specification Test Results

Boiling point 117-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR