Ethyl-2-phenyl-3-(trifluoromethyl)pyrazole-4-carboxylate

Supplier: Matrix Scientific

003888-1G 003888-5G
101786-364EA 36.92 USD
101786-364 101786-366
Ethyl-2-phenyl-3-(trifluoromethyl)pyrazole-4-carboxylate
Ethyl-2-phenyl-3-(trifluoromethyl)pyrazole-4-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₁F₃N₂O₂
MW: 284.24 g/mol
Boiling Pt: 120…125 °C (0.06 mmHg)
MDL Number: MFCD00068138
CAS Number: 112055-34-2

Order Now

Specification Test Results

Boiling point 120-125°C/0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR