Bis(pentafluorophenyl)dimethylsilane ≥97%

Supplier: Matrix Scientific

003807-5G 003807-25G
101786-272EA 129.23 USD
101786-272 101786-274
Bis(pentafluorophenyl)dimethylsilane ≥97%
Bis(pentafluorophenyl)dimethylsilane

Hazard: Irritant

Formula: C₁₄H₆F₁₀Si
Boiling Pt: 138…140 °C (14 mmHg)
MDL Number: MFCD00042066
CAS Number: 10536-62-6

Order Now

Specification Test Results

Boiling point 138-140°C/1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR