N-[3-(Trifluoromethyl)pyrid-2-yl]-N-methyl-hydrazine
Supplier: Matrix Scientific
Synonyms:
2-(N-Methylhydrazino)-3-(trifluoromethyl)pyridine
003759-1G
003759-5G
101786-184EA
89.23
USD
101786-184
101786-186
N-[3-(Trifluoromethyl)pyrid-2-yl]-N-methyl-hydrazine
N-[3-(Trifluoromethyl)pyrid-2-yl]-N-methyl-hydrazine
Hazard: Irritant
Formula:
C₇H₈F₃N₃ Boiling Pt: 90 °C (15 mmHg) |
MDL Number:
MFCD00052298 CAS Number: 175205-68-2 |
Specification Test Results
Boiling point | 90°C/15mm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...