5-(3-(Trifluoromethyl)phenyl)oxazole
Supplier: Matrix Scientific
003758-250MG
003758-1G
101786-180EA
112.31
USD
101786-180
101786-182
5-(3-(Trifluoromethyl)phenyl)oxazole
5-(3-(Trifluoromethyl)phenyl)oxazole
Hazard: Irritant
Formula:
C₁₀H₆F₃NO MW: 213.1600 g/mol Boiling Pt: 78°C/0.6mm |
MDL Number:
MFCD00085180 CAS Number: 175205-48-8 |
Specification Test Results
Boiling point | 78°C/0.6mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...