1-[4-(Trifluoromethyl)phenyl]but-1-en-3-one

Supplier: Matrix Scientific

003755-250MG 003755-1G
101786-172EA 36.92 USD
101786-172 101786-174
1-[4-(Trifluoromethyl)phenyl]but-1-en-3-one
1-[4-(Trifluoromethyl)phenyl]but-1-en-3-one

Hazard: Irritant

Formula: C₁₁H₉F₃O
Boiling Pt: 118 °C (4 mmHg)
MDL Number: MFCD00052848
CAS Number: 80992-93-4

Order Now

Specification Test Results

Boiling point 118°C/4mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR