1-[3-(Trifluoromethyl)pyridin-2-yl]piperazine

Supplier: Matrix Scientific

Synonyms: 2-(Piperazin-1-yl)-3-(trifluoromethyl)pyridine, 1-(3-(Trifluoromethyl)pyridin-2-yl)piperazine

003752-1G 003752-5G
101786-164EA 50.77 USD
101786-164 101786-166
1-[3-(Trifluoromethyl)pyridin-2-yl]piperazine
1-[3-(Trifluoromethyl)pyridin-2-yl]piperazine

Hazard: Irritant

Formula: C₁₀H₁₂F₃N₃
Boiling Pt: 106 °C (1.5 mmHg)
MDL Number: MFCD00114705
CAS Number: 87394-63-6

Order Now

Specification Test Results

Boiling point 106°C/1.5mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR