1H,1H-Perfluoro-3,7-dimethyl-1-octanol ≥97%
Supplier: Matrix Scientific
003578-5G
003578-25G
101785-886EA
153.85
USD
101785-886
101785-888
1H,1H-Perfluoro-3,7-dimethyl-1-octanol ≥97%
1H,1H-Perfluoro-3,7-dimethyl-1-octanol
Hazard: Irritant
Formula:
C₁₀H₃F₁₉O MW: 500.1 g/mol Boiling Pt: 90°C/0.1mm |
MDL Number:
MFCD01320739 CAS Number: 232587-50-7 |
Specification Test Results
Boiling point | 90°C/0.1mm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...