Ethyl-2-methyl-3-hydroxy-4,4,4-trifluorobutyrate ≥95%

Supplier: Matrix Scientific

003394-1G 003394-5G 003394-25G
101785-660EA 533.85 USD
101785-660 101785-662 101785-664
Ethyl-2-methyl-3-hydroxy-4,4,4-trifluorobutyrate ≥95%
Ethyl-2-methyl-3-hydroxy-4,4,4-trifluorobutyrate

Hazard: Irritant

Formula: C₇H₁₁F₃O₃
Boiling Pt: 180-184°C
MDL Number: MFCD00190634
CAS Number: 91600-33-8

Order Now

Specification Test Results

Boiling point 180-184°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR