Difluoromethyl-2,2,2-trifluoroethyl ether ≥97%
Supplier: Matrix Scientific
003376-5G
003376-25G
101785-634EA
287.69
USD
101785-634
101785-636
Difluoromethyl-2,2,2-trifluoroethyl ether ≥97%
Difluoromethyl-2,2,2-trifluoroethyl ether
Hazard: Irritant
Formula:
C₃H₃F₅O Boiling Pt: 28…30 °C |
MDL Number:
MFCD00153127 CAS Number: 1885-48-9 |
Specification Test Results
Boiling point | 28-30°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...