4-Fluoropentylbenzene ≥97%

Supplier: Matrix Scientific

003199-1G 003199-5G
101785-394EA 100 USD
101785-394 101785-396
4-Fluoropentylbenzene ≥97%
4-Fluoropentylbenzene

Hazard: Irritant

Formula: C₁₁H₁₅F
MW: 166.24 g/mol
Boiling Pt: 105 °C (20 mmHg)
MDL Number: MFCD00079760
CAS Number: 28593-14-8

Order Now

Specification Test Results

Boiling point 105°C/20mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR