1,2,4-Trichloro-1,1,2-trifluorobutane ≥97%

Supplier: Matrix Scientific

003080-25G 003080-100G
101785-172EA 138.46 USD
101785-172 101785-174
1,2,4-Trichloro-1,1,2-trifluorobutane ≥97%
1,2,4-Trichloro-1,1,2-trifluorobutane

Hazard: Irritant

Formula: C₄H₄Cl₃F₃
Boiling Pt: 134…136 °C
MDL Number: MFCD00077597
CAS Number: 261760-97-8

Order Now

Specification Test Results

Boiling point 134-136°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR