2,6-Dichloro-4-methylnicotinamide

Supplier: Matrix Scientific

009307-1G 009307-5G
101796-828EA 98.46 USD
101796-828 101796-830
2,6-Dichloro-4-methylnicotinamide
2,6-Dichloro-4-methylnicotinamide

Hazard: Irritant

Formula: C₇H₆Cl₂N₂O
Boiling Pt: 170…171 °C
MDL Number: MFCD00052636
CAS Number: 38841-54-2

Order Now

Specification Test Results

Boiling point 129-132°C/7


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR