1,1,1,3-Tetrafluoropropane ≥97%
Supplier: Matrix Scientific
Synonyms:
FC-254fb
009166-5G
009166-25G
101796-550EA
553.85
USD
101796-550
101796-552
1,1,1,3-Tetrafluoropropane ≥97%
1,1,1,3-Tetrafluoropropane
Hazard: Flammable Gas
Formula:
C₃H₄F₄ Boiling Pt: 29…30 °C Density: 1.2584 |
CAS Number:
460-36-6 |
Specification Test Results
Boiling point | 29-30°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...