1H,1H,11H-Perfluoroundecyl methacrylate ≥95%

Supplier: Matrix Scientific

Synonyms: 1H,1H,11H-Icosafluoroundecyl methacrylate

009145-5G 009145-25G
101796-478EA 106.15 USD
101796-478 101796-480
1H,1H,11H-Perfluoroundecyl methacrylate ≥95%
1H,1H,11H-Perfluoroundecyl methacrylate

Hazard: Irritant/Refrigerate

Formula: C₁₅H₈F₂₀O₂
MW: 600.2 g/mol
Boiling Pt: 110 °C (0.05 mmHg)
MDL Number: MFCD00042111
CAS Number: 41123-44-8

Order Now

Specification Test Results

Boiling point 110°C/0.05m

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR