Perfluoro-2,5,8,11,14,17-hexamethyl-3,6,9,12,15,18-hexaoxaheneicosanoyl fluoride ≥90%

Supplier: Matrix Scientific

009107-1G 009107-5G
101796-364EA 80 USD
101796-364 101796-366
Perfluoro-2,5,8,11,14,17-hexamethyl-3,6,9,12,15,18-hexaoxaheneicosanoyl fluoride ≥90%
Perfluoro-2,5,8,11,14,17-hexamethyl-3,6,9,12,15,18-hexaoxaheneicosanoyl fluoride

Hazard: Corrosive/Irritant

Formula: C₂₁F₄₂O₇
MW: 1162.16 g/mol
Boiling Pt: 100…102 °C (0.9 mmHg)
Density: 1.8
MDL Number: MFCD03094123
CAS Number: 13140-24-4

Order Now

Specification Test Results

Boiling point 100-102°C/0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR