2-Chloro-3-fluoro-4-(trifluoromethyl)pyridine ≥98%

Supplier: Matrix Scientific

009057-1G 009057-5G
101796-232EA 60 USD
101796-232 101796-234
2-Chloro-3-fluoro-4-(trifluoromethyl)pyridine ≥98%
2-Chloro-3-fluoro-4-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₂ClF₄N
Boiling Pt: 35-37°C/25mm
MDL Number: MFCD04038326
CAS Number: 628692-22-8

Order Now

Specification Test Results

Boiling point 35-37°C/25m

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR