Methyl-4-(trifluoromethyl)pyridine-3-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 4-(trifluoromethyl)nicotinate

009041-100MG 009041-250MG
101796-180EA 107.69 USD
101796-180 101796-182
Methyl-4-(trifluoromethyl)pyridine-3-carboxylate ≥97%
Methyl-4-(trifluoromethyl)pyridine-3-carboxylate

Hazard: Irritant

Formula: C₈H₆F₃NO₂
MW: 205.14 g/mol
Boiling Pt: 100 °C (15 mmHg)
MDL Number: MFCD00101568
CAS Number: 175204-82-7

Order Now

Specification Test Results

Boiling point 100°C/15mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR