Methyl-2-bromo-3,3,3-trifluoropropanoate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2-bromo-3,3,3-trifluoropropanoate

009025-1G 009025-5G
101796-142EA 229.23 USD
101796-142 101796-144
Methyl-2-bromo-3,3,3-trifluoropropanoate ≥97%
Methyl-2-bromo-3,3,3-trifluoropropanoate

Hazard: Irritant/Refrigerate

Formula: C₄H₄BrF₃O₂
MW: 220.9726 g/mol
Boiling Pt: 126 °C
MDL Number: MFCD04038319
CAS Number: 113816-36-7

Order Now

Specification Test Results

Boiling point 126°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR