1,1,2,3,3,3-Hexafluoropropyl difluoromethyl ether ≥98%

Supplier: Matrix Scientific

008997-1G 008997-5G
101796-052EA 130.77 USD
101796-052 101796-054
1,1,2,3,3,3-Hexafluoropropyl difluoromethyl ether ≥98%
1,1,2,3,3,3-Hexafluoropropyl difluoromethyl ether

Hazard: Irritant/Refrigerate

Formula: C₄H₂F₈O
Boiling Pt: 47 °C
MDL Number: MFCD03094220
CAS Number: 56860-85-6

Order Now

Specification Test Results

Boiling point 47°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR