1,1,2,3,3,3-Hexafluoropropyl chlorofluoromethyl ether ≥97%

Supplier: Matrix Scientific

008995-1G 008995-5G
101796-044EA 67.69 USD
101796-044 101796-046
1,1,2,3,3,3-Hexafluoropropyl chlorofluoromethyl ether ≥97%
1,1,2,3,3,3-Hexafluoropropyl chlorofluoromethyl ether

Hazard: Irritant

Formula: C₄H₂ClF₇O
Boiling Pt: 70…71 °C
MDL Number: MFCD03094548
CAS Number: 56860-86-7

Order Now

Specification Test Results

Boiling point 70-71°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR