4,4,5,5,6,6,6-Heptafluoro-2-hexene ≥97%
Supplier: Matrix Scientific
008983-5G
008983-25G
101796-016EA
138.46
USD
101796-016
101796-018
4,4,5,5,6,6,6-Heptafluoro-2-hexene ≥97%
4,4,5,5,6,6,6-Heptafluoro-2-hexene
Hazard: Irritant
Formula:
C₆H₅F₇ MW: 210.1 g/mol Boiling Pt: 62°C/743mm |
MDL Number:
MFCD00155827 CAS Number: 355-95-3 |
Specification Test Results
Boiling point | 62°C/743mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...