Diethyl tetrafluorosuccinate ≥97%

Supplier: Matrix Scientific

008912-1G 008912-5G
101795-804EA 36.92 USD
101795-804 101795-806
Diethyl tetrafluorosuccinate ≥97%
Diethyl tetrafluorosuccinate

Hazard: Irritant

Formula: C₈H₁₀F₄O₄
Boiling Pt: 78 °C (5 mmHg)
Density: 1.264
MDL Number: MFCD00015155
CAS Number: 377-71-9

Order Now

Specification Test Results

Boiling point 78°C/5mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR