3,3-Dichloro-1,1,1-trifluoropropane ≥98%

Supplier: Matrix Scientific

008911-1G 008911-5G
101795-800EA 63.08 USD
101795-800 101795-802
3,3-Dichloro-1,1,1-trifluoropropane ≥98%
3,3-Dichloro-1,1,1-trifluoropropane

Hazard: Ozone Depleter/Irritant

Formula: C₃H₃Cl₂F₃
Boiling Pt: 71…72 °C
Density: 1.4408
MDL Number: MFCD00800977
CAS Number: 460-69-5

Order Now

Specification Test Results

Boiling point 71-72°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR