3,3-Dichloro-1,1,1-trifluoroacetone hydrate ≥97%

Supplier: Matrix Scientific

008910-1G 008910-5G
101795-796EA 35.38 USD
101795-796 101795-798
3,3-Dichloro-1,1,1-trifluoroacetone hydrate ≥97%
3,3-Dichloro-1,1,1-trifluoroacetone hydrate

Hazard: Irritant

Formula: C₃H₃Cl₂F₃O₂
Boiling Pt: 103°C
MDL Number: MFCD04038304
CAS Number: 1049731-87-4

Order Now

Specification Test Results

Boiling point 103°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR