1,4-Dichlorooctafluorobutane ≥99%
Supplier: Matrix Scientific
008902-1G
008902-5G
101795-768EA
112.31
USD
101795-768
101795-770
1,4-Dichlorooctafluorobutane ≥99%
1,4-Dichlorooctafluorobutane
Hazard: Irritant
Formula:
ClF₂C(CF₂)₂CF₂Cl Boiling Pt: 64…65.5 °C |
CAS Number:
355-24-8 |
Specification Test Results
Boiling point | 64-65.5°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...