2-Bromo-3,3,4,4,4-pentafluoro-1-butene ≥98%

Supplier: Matrix Scientific

008844-1G 008844-5G
101795-588EA 150.77 USD
101795-588 101795-590
2-Bromo-3,3,4,4,4-pentafluoro-1-butene ≥98%
2-Bromo-3,3,4,4,4-pentafluoro-1-butene

Hazard: Irritant

Formula: C₄H₂BrF₅
MW: 224.96 g/mol
Boiling Pt: 56…58 °C
MDL Number: MFCD03094107
CAS Number: 68318-95-6

Order Now

Specification Test Results

Boiling point 56-58°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR