2-Butyl-1,3-benzoxazol-5-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-2-(but-1-yl)-1,3-benzoxazole

045506-500MG 045506-1G 045506-5G
101871-196EA 375.38 USD
101871-196 101871-198 101871-200
2-Butyl-1,3-benzoxazol-5-amine ≥95%
2-Butyl-1,3-benzoxazol-5-amine

Hazard: Irritant

Formula: C₁₁H₁₄N₂O
Boiling Pt: 155 °C (0.4 mmHg)
CAS Number: 885949-91-7

Order Now

Specification Test Results

Boiling point 155°C/0.4mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR