2-Chloro-4-fluoro-5-(1H-pyrrol-1-yl)benzenol ≥95%

Supplier: Matrix Scientific

045406-500MG 045406-1G 045406-5G
101870-668EA 921.54 USD
101870-668 101870-670 101870-672
2-Chloro-4-fluoro-5-(1H-pyrrol-1-yl)benzenol ≥95%
2-Chloro-4-fluoro-5-(1H-pyrrol-1-yl)benzenol

Hazard: Irritant

Formula: C₁₀H₇ClFNO
Boiling Pt: 125°C/0.5mm
MDL Number: MFCD03425807
CAS Number: 439095-63-3

Order Now

Specification Test Results

Boiling point 125°C/0.5mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR