(2-Chloro-1,3-thiazol-5-yl)methanol ≥95%
Supplier: Matrix Scientific
045339-500MG
045339-1G
045339-5G
101870-398EA
138.46
USD
101870-398
101870-400
101870-402
(2-Chloro-1,3-thiazol-5-yl)methanol ≥95%
(2-Chloro-1,3-thiazol-5-yl)methanol
Hazard: Irritant/Refrigerate
Formula:
C₄H₄NOClS Boiling Pt: 96°C/0.2mm |
MDL Number:
MFCD03001277 CAS Number: 145015-15-2 |
Specification Test Results
Boiling point | 96°C/0.2mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...