3,4-Dihydro-2H-pyran-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 3,4-Dihydro-2H-pyran-5-carboxaldehyde

045244-500MG 045244-1G 045244-5G
101869-894EA 95.38 USD
101869-894 101869-896 101869-898
3,4-Dihydro-2H-pyran-5-carbaldehyde ≥95%
3,4-Dihydro-2H-pyran-5-carbaldehyde

Hazard: Irritant

Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 84 °C (21 mmHg)
MDL Number: MFCD00085019
CAS Number: 25090-33-9

Order Now

Specification Test Results

Boiling point 84°C/21mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR