Methyl-2-methyl-2-(1H-1,2,4-triazol-1-yl)propanoate ≥95%
Supplier: Matrix Scientific
045166-500MG
045166-1G
045166-5G
101869-472EA
181.54
USD
101869-472
101869-474
101869-476
Methyl-2-methyl-2-(1H-1,2,4-triazol-1-yl)propanoate ≥95%
Methyl-2-methyl-2-(1H-1,2,4-triazol-1-yl)propanoate
Hazard: Irritant
Formula:
C₇H₁₁N₃O₂ MW: 169.18 g/mol Boiling Pt: 96 °C (1 mmHg) |
MDL Number:
MFCD06659569 CAS Number: 885949-78-0 |
Specification Test Results
Boiling point | 96°C/1mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...